Learn More
3M™ Nomad™ Terra Heavy Traffic Entrance Matting 8100
Entrance coiled vinyl scraper matting designed for heavy traffic applications. The construction allows dirt and moisture to flow through the matting so it is not retracked into the building. Grease and oil resistant.
Brand:  3M™ Nomad™ 7100116624
 View more versions of this product
Product Code. 17138710
  • 304.00 € / 1 roll
Estimated Shipment: 11-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Unbacked scraper matting designed for high traffic applications
  • High quality vinyl offers excellent crush resistance
  • Unbacked, open construction of the mat allows water to drain and soil to pass through, out of sight
  • Easily vacuum out trapped dirt
TRUSTED_SUSTAINABILITY
Specifications
Specifications
152 x 91.4 cm
Nomad™ Terra Dirt Control Mats
Gray
Elastic Vinyl
Heavy traffic areas
17 mm
152 cm
91.4 cm