Learn More
tert-Butanol, 99.5%, extra pure, Thermo Scientific Chemicals
44.70 € - 606.00 €
Chemical Identifiers
CAS | 75-65-0 |
---|---|
Molecular Formula | C4H10O |
Molecular Weight (g/mol) | 74.123 |
MDL Number | MFCD00004464 |
InChI Key | DKGAVHZHDRPRBM-UHFFFAOYSA-N |
Synonym | tert-butanol, tert-butyl alcohol, 2-methyl-2-propanol, t-butanol, t-butyl hydroxide, 1,1-dimethylethanol, trimethylcarbinol, trimethyl methanol, 2-propanol, 2-methyl, t-butyl alcohol |
PubChem CID | 6386 |
ChEBI | CHEBI:45895 |
IUPAC Name | 2-methylpropan-2-ol |
SMILES | CC(C)(C)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10000730
|
Thermo Scientific Acros
107710010 |
1 L | Plastic Bottle |
44.70 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10775361
|
Thermo Scientific Acros
107710025 |
2.5 L | Plastic Bottle |
88.80 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10470364
|
Thermo Scientific Acros
107710100 |
10 L | Plastic Drum |
606.00 €
10 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
75-65-0 | |
74.123 | |
DKGAVHZHDRPRBM-UHFFFAOYSA-N | |
6386 | |
2-methylpropan-2-ol |
C4H10O | |
MFCD00004464 | |
tert-butanol, tert-butyl alcohol, 2-methyl-2-propanol, t-butanol, t-butyl hydroxide, 1,1-dimethylethanol, trimethylcarbinol, trimethyl methanol, 2-propanol, 2-methyl, t-butyl alcohol | |
CHEBI:45895 | |
CC(C)(C)O |
Specifications
25.0°C to 25.5°C | |
Colorless | |
83.0°C | |
Authentic | |
1.3860 to 1.3880 | |
0.1% max. | |
C4H10O | |
MFCD00004464 | |
01, 379 | |
15, 1544 | |
Solubility in water: soluble. Other solubilities: soluble in alcohol, ether, acetone | |
CC(C)(C)O | |
74.123 | |
CHEBI:45895 | |
99.5% | |
tert-Butanol |
75-65-0 | |
0.7800g/mL | |
11°C | |
Liquid After Melting | |
99.5% | |
0.78 | |
(CH3)3COH | |
3.35 mPa.s (30°C) | |
0.03% max. (K.F.) | |
tert-butanol, tert-butyl alcohol, 2-methyl-2-propanol, t-butanol, t-butyl hydroxide, 1,1-dimethylethanol, trimethylcarbinol, trimethyl methanol, 2-propanol, 2-methyl, t-butyl alcohol | |
DKGAVHZHDRPRBM-UHFFFAOYSA-N | |
2-methylpropan-2-ol | |
6386 | |
74.12 | |
Extra Pure |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if inhaled.
Highly flammable liquid and vapor.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
- tert-Butanol
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- Acute toxicity Category 4
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only