missing translation for 'onlineSavingsMsg'
Learn More

tert-Butanol, CHROMASOLV™, for HPLC, ≥99.5%, Honeywell Riedel-de Haën™

CHROMASOLV™, for HPLC,≥99.5%

91.65 € - 551.60 €

Chemical Identifiers

CAS 75-65-0
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.123
MDL Number MFCD00004464
InChI Key DKGAVHZHDRPRBM-UHFFFAOYSA-N
Synonym tert-butanol, tert-butyl alcohol, 2-methyl-2-propanol, t-butanol, t-butyl hydroxide, 1,1-dimethylethanol, trimethylcarbinol, trimethyl methanol, 2-propanol, 2-methyl, t-butyl alcohol
PubChem CID 6386
ChEBI CHEBI:45895
IUPAC Name 2-methylpropan-2-ol
SMILES CC(C)(C)O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15633140
View Documents Promotion Details
Honeywell-Chromasolv
308250-1L
1 L Glass Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15643140
View Documents Promotion Details
Honeywell-Chromasolv
308250-2.5L
2.5 L Glass Bottle
227.08 €
2.50 L
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15653140
View Documents Promotion Details
Honeywell-Chromasolv
308250-4X2.5L
4 x 2.5 L Glass Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency
Specifications

Chemical Identifiers

75-65-0
74.123
DKGAVHZHDRPRBM-UHFFFAOYSA-N
6386
2-methylpropan-2-ol
C4H10O
MFCD00004464
tert-butanol, tert-butyl alcohol, 2-methyl-2-propanol, t-butanol, t-butyl hydroxide, 1,1-dimethylethanol, trimethylcarbinol, trimethyl methanol, 2-propanol, 2-methyl, t-butyl alcohol
CHEBI:45895
CC(C)(C)O

Specifications

23°C to 26°C (literature)
75-65-0
0.775 g/mL at 25 °C(lit.)
11°C
(CH3)3COH
UN1120
tert-butanol, tert-butyl alcohol, 2-methyl-2-propanol, t-butanol, t-butyl hydroxide, 1,1-dimethylethanol, trimethylcarbinol, trimethyl methanol, 2-propanol, 2-methyl, t-butyl alcohol
CC(C)(C)O
74.123
CHEBI:45895
>99.5%
tert-Butanol, CHROMASOLV™, for HPLC, ≥99.5%
max. 0.01 at 350 nm, max. 0.01 at 300 nm, max. 0.20 at 250 nm, max. 0.50 at 230 nm, max. 1.00 at 215 nm
100.0000%
81°C to 83°C
C4H10O
MFCD00004464
906698
DKGAVHZHDRPRBM-UHFFFAOYSA-N
2-methylpropan-2-ol
6386
74.12
HPLC
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 720 days from date of manufacture

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Butanols

missing translation for 'einecsNumber' : 200-889-7

Recommended Storage : Room Temp

SDS
Documents

Documents

Product Certifications