Learn More
tert-Butyl 2,2,2-trichloroacetimidate, 95%, Thermo Scientific Chemicals
49.50 € - 507.00 €
Chemical Identifiers
CAS | 98946-18-0 |
---|---|
Molecular Formula | C6H10Cl3NO |
Molecular Weight (g/mol) | 218.50 |
MDL Number | MFCD00077410 |
InChI Key | CQXDYHPBXDZWBA-UHFFFAOYSA-N |
Synonym | tert-butyl 2,2,2-trichloroacetimidate, tert-butyl trichloroacetimidate, tert-butyl-2,2,2-trichloroacetimidate, tert-butyl 2,2,2-trichloroethanecarboximidate, 2,2,2-trichloroacetimidic acid tert-butyl ester, t-butyl trichloroacetimidate, t-butyl 2,2,2-trichloroacetimidate, tert-butyl2,2,2-trichloroacetimidate, ethanimidic acid, 2,2,2-trichloro-, 1,1-dimethylethyl ester, 1-tert-butoxy-2,2,2-trichloroethanimine |
PubChem CID | 2734700 |
IUPAC Name | tert-butyl 2,2,2-trichloroethanimidate |
SMILES | CC(C)(C)OC(=N)C(Cl)(Cl)Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10767403
|
Thermo Scientific Acros
369330050 |
5 g | Glass bottle |
49.50 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10415053
|
Thermo Scientific Acros
369330250 |
25 g | Glass bottle |
167.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11905601
|
Thermo Scientific Acros
369331000 |
100 g | Glass Bottle |
507.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagentChemical Identifiers
98946-18-0 | |
218.50 | |
CQXDYHPBXDZWBA-UHFFFAOYSA-N | |
2734700 | |
CC(C)(C)OC(=N)C(Cl)(Cl)Cl |
C6H10Cl3NO | |
MFCD00077410 | |
tert-butyl 2,2,2-trichloroacetimidate, tert-butyl trichloroacetimidate, tert-butyl-2,2,2-trichloroacetimidate, tert-butyl 2,2,2-trichloroethanecarboximidate, 2,2,2-trichloroacetimidic acid tert-butyl ester, t-butyl trichloroacetimidate, t-butyl 2,2,2-trichloroacetimidate, tert-butyl2,2,2-trichloroacetimidate, ethanimidic acid, 2,2,2-trichloro-, 1,1-dimethylethyl ester, 1-tert-butoxy-2,2,2-trichloroethanimine | |
tert-butyl 2,2,2-trichloroethanimidate |
Specifications
98946-18-0 | |
Colorless to Yellow | |
64.0°C to 66.0°C (15.0 mmHg) | |
Authentic | |
C6H10Cl3NO | |
1.4550 to 1.4570 | |
1.222 | |
CQXDYHPBXDZWBA-UHFFFAOYSA-N | |
tert-butyl 2,2,2-trichloroethanimidate | |
2734700 | |
95% | |
tert-Butyl 2, 2, 2-trichloroacetimidate |
20.0°C | |
1.2220g/mL | |
55°C | |
94% min. (GC) | |
CCl3C(=NH)OC(CH3)3 | |
MFCD00077410 | |
tert-butyl 2,2,2-trichloroacetimidate, tert-butyl trichloroacetimidate, tert-butyl-2,2,2-trichloroacetimidate, tert-butyl 2,2,2-trichloroethanecarboximidate, 2,2,2-trichloroacetimidic acid tert-butyl ester, t-butyl trichloroacetimidate, t-butyl 2,2,2-trichloroacetimidate, tert-butyl2,2,2-trichloroacetimidate, ethanimidic acid, 2,2,2-trichloro-, 1,1-dimethylethyl ester, 1-tert-butoxy-2,2,2-trichloroethanimine | |
CC(C)(C)OC(=N)C(Cl)(Cl)Cl | |
218.50 | |
218.51 | |
Liquid or Low Melting Crystalline Mass |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
- tert-Butyl 2,2,2-trichloroacetimidate
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.