missing translation for 'onlineSavingsMsg'
Learn More
tert-Butyl 2-amino-2-thioxoethylcarbamate, 90%, Thermo Scientific™ Product Code.: 10443245

tert-Butyl 2-amino-2-thioxoethylcarbamate, 90%, Thermo Scientific™

Product Code. 10443245
1 g, Amber Glass Bottle
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 10443245

Brand: Thermo Scientific Maybridge MO07925DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 89226-13-1
Molecular Formula C7H14N2O2S
Molecular Weight (g/mol) 190.261
MDL Number MFCD09025922
InChI Key AGBIUUFZUPNDTM-UHFFFAOYSA-N
Synonym tert-butyl 2-amino-2-thioxoethyl carbamate, tert-butyl 2-amino-2-thioxoethylcarbamate, tert-butyl n-carbamothioylmethyl carbamate, tert-butyl n-2-amino-2-sulfanylideneethyl carbamate, carbamic acid, 2-amino-2-thioxoethyl-, 1,1-dimethylethyl ester, carbamic acid,n-2-amino-2-thioxoethyl-, 1,1-dimethylethyl ester, acmc-20ljf8, n-tert-butoxycarbonylglycinethioamide, tert-butoxycarbonylamino acetothioamide, n-t-butoxycarbonylamino acetothioamide
PubChem CID 5324304
IUPAC Name tert-butyl N-(2-amino-2-sulfanylideneethyl)carbamate
SMILES CC(C)(C)OC(=O)NCC(=S)N

Specifications

CAS Min % 90.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 190.27
Percent Purity 90%
Chemical Name or Material Tert-butyl 2-amino-2-thioxoethylcarbamate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
tert-Butyl 2-amino-2-thioxoethylcarbamate, 90%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.