missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 2-aminoisobutyrate hydrochloride, 98%, Thermo Scientific Chemicals

Product Code. 15478536
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

tert-Butyl 2-aminoisobutyrate hydrochloride is used as intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl 2-aminoisobutyrate hydrochloride is used as intermediates.

Solubility
Soluble in water or 1% acetic acid.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84758-81-6
Molecular Formula C8H17NO2
Molecular Weight (g/mol) 159.23
MDL Number MFCD00237698
InChI Key LSVYCJILORYVCD-UHFFFAOYSA-N
Synonym alpha-aminoisobutyric acid t-butyl ester, h-aib-otbu.hcl, tert-butyl 2-methylalaninate, 2-aminoisobutyric acid t-butyl ester, alanine, 2-methyl-, 1,1-dimethylethyl ester, 2-amino-2-methylpropionic acid tert-butyl ester, h-aib-otbu inverted exclamation mark currencyhcl, t-butyl 2-methylalaninate, 2-methylalanine t-butyl ester, h-alpha-me-ala-otbu . hcl
PubChem CID 7019940
IUPAC Name tert-butyl 2-amino-2-methylpropanoate
SMILES CC(C)(C)OC(=O)C(C)(C)N

Specifications

Color White
Quantity 250 mg
Solubility Information Soluble in water or 1% acetic acid.
Formula Weight 195.69
Percent Purity 98%
Physical Form Powder
Chemical Name or Material tert-Butyl 2-aminoisobutyrate hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'