missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 3-oxocyclobutanecarboxylate, 97%, Thermo Scientific Chemicals

Product Code. 15418658
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 145549-76-4
Molecular Formula C9H14O3
Molecular Weight (g/mol) 170.208
MDL Number MFCD12827541
InChI Key JINYZTGTQXDUQR-UHFFFAOYSA-N
Synonym tert-butyl 3-oxocyclobutanecarboxylate, 3-oxo-cyclobutanecarboxylic acid tert-butyl ester, cyclobutanecarboxylic acid, 3-oxo-, 1,1-dimethylethyl ester, tert-butyl 3-oxo-cyclobutanecarboxylate, 1-boc-cyclobutan-3-one, t-butyl 3-oxocyclobutanecarboxylate, 3-tert-butyloxycarbonylcyclobutanone, tert-butyl3-oxocyclobutanecarboxylate, tert-butyl 3-oxocyclobutane carboxylate, t-butyl-3-oxocyclobutanecarboxylate
PubChem CID 10261520
IUPAC Name tert-butyl 3-oxocyclobutane-1-carboxylate
SMILES CC(C)(C)OC(=O)C1CC(=O)C1

Specifications

Quantity 250 mg
Formula Weight 170.2
Percent Purity 97%
Chemical Name or Material tert-Butyl 3-oxocyclobutanecarboxylate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'