missing translation for 'onlineSavingsMsg'
Learn More
tert-Butyl 4-(2-(aminomethyl)phenyl)piperazine-1-carboxylate, ≥97%, Thermo Scientific™ Product Code.: 10085822

tert-Butyl 4-(2-(aminomethyl)phenyl)piperazine-1-carboxylate, ≥97%, Thermo Scientific™

Product Code. 10085822
250 mg, Amber Glass Bottle
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 10085822

Brand: Thermo Scientific Maybridge CC30713CB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 174855-53-9
Molecular Formula C16H25N3O2
Molecular Weight (g/mol) 291.395
MDL Number MFCD04115066
InChI Key UINNZXQKTNAOBL-UHFFFAOYSA-N
Synonym tert-butyl 4-2-aminomethyl phenyl piperazine-1-carboxylate, 1-boc-4-2-aminomethyl phenyl piperazine, 1-boc-4-2-aminomethyl-phenyl-piperazine, 1-2-aminomethyl-phenyl-piperazine-4-carboxylic acid tert-butyl ester, 1-piperazinecarboxylicacid, 4-2-aminomethyl phenyl-, 1,1-dimethylethyl ester, 1-2-aminomethyl-phenyl-piperazine-4-carboxylic a, 2-4-t-butoxycarbonylpiperazin-1-yl phenylmethylamine, 4-2-aminomethyl-phenyl-piperazine-1-carboxylic acid tert-butyl ester, 4-2-aminomethyl phenyl-1-piperazinecarboxylic acid tert-butyl esterShow MoreShow Less
PubChem CID 4961274
IUPAC Name tert-butyl 4-[2-(aminomethyl)phenyl]piperazine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCN(CC1)C2=CC=CC=C2CN

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 291.39
Percent Purity 97%
Chemical Name or Material Tert-butyl 4-[2-(aminomethyl)phenyl]piperazine-1-carboxylate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
tert-Butyl 4-(2-(aminomethyl)phenyl)piperazine-1-carboxylate, ≥97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.