Learn More
tert-Butyl 4-(2-aminophenyl)tetrahydro-1(2H)-pyrazinecarboxylate, 97%, Thermo Scientific™
58.00 € - 122.00 €
Chemical Identifiers
CAS | 170017-74-0 |
---|---|
Molecular Formula | C15H23N3O2 |
Molecular Weight (g/mol) | 277.37 |
MDL Number | MFCD04115046 |
InChI Key | LNDQGWAWYKFYAO-UHFFFAOYSA-N |
Synonym | 1-boc-4-2-aminophenyl piperazine, tert-butyl 4-2-aminophenyl piperazine-1-carboxylate, 2-4-boc-piperazin-1-yl aniline, tert-butyl 4-2-aminophenyl tetrahydro-1 2h-pyrazinecarboxylate, 1-boc-4-2-aminophenyl-piperazine, 4-2-amino-phenyl-piperazine-1-carboxylic acid tert-butyl ester, 2-4-tert-butoxycarbonyl piperazin-1-yl aniline, 4-2-aminophenyl piperazine, n1-boc protected, pubchem12189, acmc-1bzzw |
PubChem CID | 2795530 |
IUPAC Name | tert-butyl 4-(2-aminophenyl)piperazine-1-carboxylate |
SMILES | CC(C)(C)OC(=O)N1CCN(CC1)C1=CC=CC=C1N |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10259202
|
Thermo Scientific Maybridge
CC30714CB |
250 mg | Amber Glass Bottle |
58.00 €
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10371013
|
Thermo Scientific Maybridge
CC30714DA |
1 g | Amber Glass Bottle |
122.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
170017-74-0 | |
277.37 | |
LNDQGWAWYKFYAO-UHFFFAOYSA-N | |
2795530 | |
CC(C)(C)OC(=O)N1CCN(CC1)C1=CC=CC=C1N |
C15H23N3O2 | |
MFCD04115046 | |
1-boc-4-2-aminophenyl piperazine, tert-butyl 4-2-aminophenyl piperazine-1-carboxylate, 2-4-boc-piperazin-1-yl aniline, tert-butyl 4-2-aminophenyl tetrahydro-1 2h-pyrazinecarboxylate, 1-boc-4-2-aminophenyl-piperazine, 4-2-amino-phenyl-piperazine-1-carboxylic acid tert-butyl ester, 2-4-tert-butoxycarbonyl piperazin-1-yl aniline, 4-2-aminophenyl piperazine, n1-boc protected, pubchem12189, acmc-1bzzw | |
tert-butyl 4-(2-aminophenyl)piperazine-1-carboxylate |
Specifications
170017-74-0 | |
100.0 | |
C15H23N3O2 | |
1-boc-4-2-aminophenyl piperazine, tert-butyl 4-2-aminophenyl piperazine-1-carboxylate, 2-4-boc-piperazin-1-yl aniline, tert-butyl 4-2-aminophenyl tetrahydro-1 2h-pyrazinecarboxylate, 1-boc-4-2-aminophenyl-piperazine, 4-2-amino-phenyl-piperazine-1-carboxylic acid tert-butyl ester, 2-4-tert-butoxycarbonyl piperazin-1-yl aniline, 4-2-aminophenyl piperazine, n1-boc protected, pubchem12189, acmc-1bzzw | |
CC(C)(C)OC(=O)N1CCN(CC1)C1=CC=CC=C1N | |
277.37 | |
277.37 | |
Tert-butyl 4-(2-aminophenyl)tetrahydro-1(2H)-pyrazinecarboxylate |
96.5 | |
97% | |
MFCD04115046 | |
LNDQGWAWYKFYAO-UHFFFAOYSA-N | |
tert-butyl 4-(2-aminophenyl)piperazine-1-carboxylate | |
2795530 | |
97% |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
Your input is important to us. Please complete this form to provide feedback related to the content on this product.