missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,2,3,6-tetrahydropyridine-1-carboxylate, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
10 g
Specifications
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 5 g |
Formula Weight | 309.21 |
Percent Purity | 97% |
Chemical Name or Material | N-(tert-Butoxycarbonyl)-3, 6-dihydro-2H-pyridine-4-boronic acidpinacol ester |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
tert-Butyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,2,3,6-tetrahydropyridine-1-carboxylate, 97%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction