missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 4-{4-[(methylamino)methyl]pyrid-2-yl}piperazine-1-carboxylate, 97%, Thermo Scientific™

Product Code. 10377494
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 946409-15-0
Molecular Formula C16H26N4O2
Molecular Weight (g/mol) 306.41
MDL Number MFCD09966148
InChI Key BNLNDKXJNWJYKP-UHFFFAOYSA-N
Synonym tert-butyl 4-4-methylamino methyl pyridin-2-yl piperazine-1-carboxylate, tert-butyl 4-4-methylamino methyl pyrid-2-yl piperazine-1-carboxylate
PubChem CID 28765107
IUPAC Name tert-butyl 4-[4-(methylaminomethyl)pyridin-2-yl]piperazine-1-carboxylate
SMILES CNCC1=CC(=NC=C1)N1CCN(CC1)C(=O)OC(C)(C)C

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 306.41
Percent Purity 97%
Chemical Name or Material tert-Butyl 4-{4-[(methylamino)methyl]pyrid-2-yl}piperazine-1-carboxylate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'