missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 4-[5-(hydroxymethyl)pyrid-2-yl]piperazine-1-carboxylate, ≥97%, Thermo Scientific™

Product Code. 10632065
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 857284-20-9
Molecular Formula C15H23N3O3
Molecular Weight (g/mol) 293.367
MDL Number MFCD08060526
InChI Key QKCHWJYIPFBVRD-UHFFFAOYSA-N
Synonym tert-butyl 4-5-hydroxymethyl pyrid-2-yl piperazine-1-carboxylate, tert-butyl 4-5-hydroxymethyl pyridin-2-yl piperazine-1-carboxylate, 1-piperazinecarboxylicacid, 4-5-hydroxymethyl-2-pyridinyl-, 1,1-dimethylethyl ester, 1-piperazinecarboxylicacid,4-5-hydroxymethyl-2-pyridinyl-,1,1-dimethylethyl ester
PubChem CID 7537576
IUPAC Name tert-butyl 4-[5-(hydroxymethyl)pyridin-2-yl]piperazine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCN(CC1)C2=NC=C(C=C2)CO

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 293.36
Percent Purity 97%
Chemical Name or Material tert-Butyl 4-[5-(hydroxymethyl)pyrid-2-yl]piperazine-1-carboxylate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'