missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butyl 6-fluoro-1H-pyrazolo[3,4-b]pyridine-3-carboxylate, ≥97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Formula Weight | 237.23 |
Percent Purity | 97% |
Chemical Name or Material | tert-Butyl 6-fluoro-1H-pyrazolo[3, 4-b]pyridine-3-carboxylate |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 7
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
tert-Butyl 6-fluoro-1H-pyrazolo[3,4-b]pyridine-3-carboxylate, ≥97%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction