Learn More
tert-Butyl bromoacetate, 98%, Thermo Scientific Chemicals
Description
tert-Butyl bromoacetate is used as an intermediate in the production of dyes, pharmaceuticals, agrochemicals and in organic compounds. It finds application as an alkylating agent. It plays an important role to prepare (2-oxo-4-vinyl-azetidin-1-yl)-acetic acid tert-butyl ester and in collagenase inhibitors synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
tert-Butyl bromoacetate is used as an intermediate in the production of dyes, pharmaceuticals, agrochemicals and in organic compounds. It finds application as an alkylating agent. It plays an important role to prepare (2-oxo-4-vinyl-azetidin-1-yl)-acetic acid tert-butyl ester and in collagenase inhibitors synthesis.
Solubility
Miscible with ethanol, chloroform and ethyl acetate. Immiscible with water.
Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong bases.

Specifications
Specifications
Density | 1.33 |
Boiling Point | 175°C |
Flash Point | 49°C (120°F) |
Odor | Pungent |
Linear Formula | BrCH2CO2C(CH3)3 |
Refractive Index | 1.445 |
Quantity | 250 g |
UN Number | UN2924 |
Beilstein | 1753010 |
Solubility Information | Miscible with ethanol,chloroform and ethyl acetate. Immiscible with water. |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10222734 | Certificate of Analysis | 30/08/2024 | A14917.09,A14917.18,A14917.30 |
10224185 | Certificate of Analysis | 30/08/2024 | A14917.09,A14917.18,A14917.30 |
J26P04 | Certificate of Analysis | 22/11/2021 | A14917.09,A14917.18,A14917.30 |
10146160 | Certificate of Analysis | 22/11/2021 | A14917.09,A14917.18,A14917.30 |
10135777 | Certificate of Analysis | 22/11/2021 | A14917.09,A14917.18,A14917.30 |
Safety and Handling



- tert-Butyl bromoacetate
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 B
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : tert-Butyl bromoacetate
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.