missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butyl carbamate, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
tert-Butyl carbamate is used in palladium-catalyzed synthesis of N-Boc-protected anilines. It was used in the synthesis of tetrasubstituted pyrroles, functionalized with ester or ketone groups at C-3 position.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
tert-Butyl carbamate is used in palladium-catalyzed synthesis of N-Boc-protected anilines. It was used in the synthesis of tetrasubstituted pyrroles, functionalized with ester or ketone groups at C-3 position.
Solubility
Soluble in methylene chloride, chloroform, and alcohols. Slightly soluble in petroleum ether and water.
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
tert-Butyl carbamate is used in palladium-catalyzed synthesis of N-Boc-protected anilines. It was used in the synthesis of tetrasubstituted pyrroles, functionalized with ester or ketone groups at C-3 position.
Solubility
Soluble in methylene chloride, chloroform, and alcohols. Slightly soluble in petroleum ether and water.
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Specifications
Specifications
Melting Point | 106°C to 110°C |
Beilstein | 1744500 |
Solubility Information | Soluble in methylene chloride,chloroform,and alcohols. Slightly soluble in petroleum ether and water. |
Formula Weight | 117.15 |
Percent Purity | ≥98% |
Quantity | 1 g |
Chemical Name or Material | tert-Butyl carbamate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'