missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl chloride, 98+%, Thermo Scientific Chemicals

Product Code. 11309659
Click to view available options
Quantity:
250 mL
500 mL
1000 mL
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

tert-Butyl chloride plays an important role as a starting material to perform nucleophilic substitution reactions in order to prepare alcohol and alkoxides salts. It is used as an alkylating agent for the introduction of tert-butyl group and is also involved in Friedel-Crafts reactions. It is employed as an intermediate for the synthesis of agrochemicals and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl chloride plays an important role as a starting material to perform nucleophilic substitution reactions in order to prepare alcohol and alkoxides salts. It is used as an alkylating agent for the introduction of tert-butyl group and is also involved in Friedel-Crafts reactions. It is employed as an intermediate for the synthesis of agrochemicals and pharmaceuticals.

Solubility
Miscible with alcohol, benzene, chloroform and ether. Slightly miscible with water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 507-20-0
Molecular Formula C4H9Cl
Molecular Weight (g/mol) 92.566
MDL Number MFCD00000816
InChI Key NBRKLOOSMBRFMH-UHFFFAOYSA-N
Synonym tert-butyl chloride, propane, 2-chloro-2-methyl, tert-butylchloride, chlorotrimethylmethane, trimethylchloromethane, 2-chloroisobutane, t-butyl chloride, t-butylchloride, 2-methyl-2-chloropropane, 2-methyl-2-propyl chloride
PubChem CID 10486
IUPAC Name 2-chloro-2-methylpropane
SMILES CC(C)(C)Cl

Specifications

Melting Point -26°C
Density 0.846
Boiling Point 50°C to 52°C
Flash Point −27°C (−16°F)
Odor Mild
Refractive Index 1.3848
Quantity 500 mL
UN Number UN1127
Beilstein 1730872
Merck Index 14,1562
Solubility Information Miscible with alcohol,benzene,chloroform and ether. Slightly miscible with water.
Formula Weight 92.57
Percent Purity ≥98%
Chemical Name or Material tert-Butyl chloride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.