missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butyl chloride, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mL
500 mL
1000 mL
Description
tert-Butyl chloride plays an important role as a starting material to perform nucleophilic substitution reactions in order to prepare alcohol and alkoxides salts. It is used as an alkylating agent for the introduction of tert-butyl group and is also involved in Friedel-Crafts reactions. It is employed as an intermediate for the synthesis of agrochemicals and pharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
tert-Butyl chloride plays an important role as a starting material to perform nucleophilic substitution reactions in order to prepare alcohol and alkoxides salts. It is used as an alkylating agent for the introduction of tert-butyl group and is also involved in Friedel-Crafts reactions. It is employed as an intermediate for the synthesis of agrochemicals and pharmaceuticals.
Solubility
Miscible with alcohol, benzene, chloroform and ether. Slightly miscible with water.
Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong bases.
tert-Butyl chloride plays an important role as a starting material to perform nucleophilic substitution reactions in order to prepare alcohol and alkoxides salts. It is used as an alkylating agent for the introduction of tert-butyl group and is also involved in Friedel-Crafts reactions. It is employed as an intermediate for the synthesis of agrochemicals and pharmaceuticals.
Solubility
Miscible with alcohol, benzene, chloroform and ether. Slightly miscible with water.
Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong bases.
Specifications
Melting Point | -26°C |
Density | 0.846 |
Boiling Point | 50°C to 52°C |
Flash Point | −27°C (−16°F) |
Odor | Mild |
Refractive Index | 1.3848 |
Quantity | 250 mL |
UN Number | UN1127 |
Beilstein | 1730872 |
Merck Index | 14,1562 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'