missing translation for 'onlineSavingsMsg'
Learn More
tert-Butyl chloroacetate, 98%, Thermo Scientific Chemicals Product Code.: 11442518

tert-Butyl chloroacetate, 98%, Thermo Scientific Chemicals

Product Code. 11442518
100 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11442518

Brand: Thermo Scientific Alfa Aesar A15833.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

tert-Butyl chloroacetate is used in the synthesis of imidazol-1-yl-acetic acid hydrochloride, cis-disubstituted aziridine ester via aza-Darzens reaction and 1,10-diaza-18-crown-6 based sensors bearing a coumarin fluorophore.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl chloroacetate is used in the synthesis of imidazol-1-yl-acetic acid hydrochloride, cis-disubstituted aziridine ester via aza-Darzens reaction and 1,10-diaza-18-crown-6 based sensors bearing a coumarin fluorophore.

Solubility
Decomposes in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Moisture sensitive. Incompatible with strong bases and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-59-5
Molecular Formula C6H11ClO2
Molecular Weight (g/mol) 150.602
MDL Number MFCD00000930
InChI Key KUYMVWXKHQSIAS-UHFFFAOYSA-N
Synonym tert-butyl chloroacetate, chloroacetic acid tert-butyl ester, acetic acid, chloro-, 1,1-dimethylethyl ester, t-butyl chloroacetate, tert-butylchloroacetate, acetic acid, chloro-, tert-butyl ester, chloroacetic acid t-butyl ester, acetic acid, 2-chloro-, 1,1-dimethylethyl ester, t-butylchloroacetate, t-butyl-chloroacetate
PubChem CID 66052
IUPAC Name tert-butyl 2-chloroacetate
SMILES CC(C)(C)OC(=O)CCl

Specifications

Density 1.053
Boiling Point 155°C
Flash Point 46°C (115°F)
Odor Ester-like
Linear Formula ClCH2CO2C(CH3)3
Refractive Index 1.423
Quantity 100 g
UN Number UN2929
Beilstein 1753006
Merck Index 14,1563
Solubility Information Decomposes in water.
Formula Weight 150.61
Percent Purity 98%
Chemical Name or Material tert-Butyl chloroacetate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10249895 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog NumberA15833.22,A15833.14,A15833.36
Lot Number10204705 Certificate TypeCertificate of Analysis Date25/07/2022 Catalog NumberA15833.22,A15833.14,A15833.36
Lot Number10240205 Certificate TypeCertificate of Analysis Date20/04/2022 Catalog NumberA15833.22,A15833.14,A15833.36
Lot Number10148743 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA15833.22,A15833.14,A15833.36
Lot Number10153579 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA15833.22,A15833.14,A15833.36
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H331-Toxic if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P311-Call a POISON CENTER/doctor/ .

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
tert-Butyl chloroacetate, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.