missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl hydroperoxide, 70% Solution in water, Thermo Scientific Chemicals

Product Code. 10703571
Click to view available options
Quantity:
5 g
250 g
500 g
2.5 kg
10 kg
Packaging:
Plastic Can
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-91-2, 7732-18-5
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.12
InChI Key CIHOLLKRGTVIJN-UHFFFAOYSA-N
Synonym tert-butyl hydroperoxide, tbhp, t-butyl hydroperoxide, tert-butylhydroperoxide, perbutyl h, t-butylhydroperoxide, cadox tbh, hydroperoxide, 1,1-dimethylethyl, 1,1-dimethylethyl hydroperoxide, terc. butylhydroperoxid
PubChem CID 6410
ChEBI CHEBI:64090
IUPAC Name 2-hydroperoxy-2-methylpropane
SMILES CC(C)(C)OO

Specifications

Melting Point -2.8°C
Color Colorless
Density 0.9400g/mL
Boiling Point 37.0°C (15.0 mmHg)
Flash Point 43°C
Packaging Plastic bottle
Linear Formula (CH3)3COOH
Refractive Index 1.3860 to 1.3880
Quantity 5 g
Beilstein 01,IV,1616
Fieser 01,88; 02,49; 03,37; 05,75; 06,81; 07,43; 08,62; 09,78; 10,64; 11,91; 12,88; 14,60; 15,55; 16,53; 17,56
Merck Index 15,1573
Specific Gravity 0.94
Solubility Information Solubility in water: miscible. Other solubilities: soluble in hydroxide solutions,soluble in most common organic solvents
Viscosity 4.1 mPa.s (20°C)
Formula Weight 90.12
Physical Form Liquid
Chemical Name or Material tert-Butyl hydroperoxide
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • tert-Butyl hydroperoxide, 70% solution in water
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Germ cell mutagenicity Category 2
  • Organic peroxide Type E & F
  • Skin sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • H341-Suspected of causing genetic defects.
  • H411-Toxic to aquatic life with long lasting effects.
  • H311-Toxic in contact with skin.
  • H302-Harmful if swallowed.
  • H242-Heating may cause a fire.
  • H226-Flammable liquid and vapour.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'