missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butyl methyl ether, 99+%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
1 L
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | -110.0°C |
CAS Min % | 99 |
Color | Colorless |
CAS Max % | 100 |
Density | 0.7404g/mL |
Boiling Point | 54.0°C to 56.0°C |
Quantity | 1 L |
Flash Point | −28°C |
Infrared Spectrum | Authentic |
Packaging | AcroSeal™ Glass Bottle |
Show More |
Safety and Handling
Product Identifier
- tert-Butyl methyl ether
- Danger
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- H225-Highly flammable liquid and vapour.
- H315-Causes skin irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'