Learn More
tert-Butyl methyl ether, 99%, pure, Thermo Scientific Chemicals
21.11 € - 409.04 €
Chemical Identifiers
CAS | 1634-04-4 |
---|---|
Molecular Formula | C5H12O |
Molecular Weight (g/mol) | 88.15 |
MDL Number | MFCD00008812 |
InChI Key | BZLVMXJERCGZMT-UHFFFAOYSA-N |
Synonym | tert-butyl methyl ether, methyl tert-butyl ether, mtbe, methyl t-butyl ether, t-butyl methyl ether, methyl tertiary-butyl ether, propane, 2-methoxy-2-methyl, methyl-tert-butyl ether, methyl-t-butyl ether, 2-methyl-2-methoxypropane |
PubChem CID | 15413 |
ChEBI | CHEBI:27642 |
IUPAC Name | 2-methoxy-2-methylpropane |
SMILES | CC(C)(C)OC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10772601
|
Thermo Scientific Acros
177042500 |
250 mL | Glass Bottle |
21.11 €
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10664672
|
Thermo Scientific Acros
177040010 |
1 L | Glass Bottle |
36.98 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10522011
|
Thermo Scientific Acros
177040025 |
2.5 L | Glass Bottle |
63.82 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10501422
|
Thermo Scientific Acros
177040250 |
25 L | Metal Drum |
409.04 €
25 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1634-04-4 | |
88.15 | |
BZLVMXJERCGZMT-UHFFFAOYSA-N | |
15413 | |
2-methoxy-2-methylpropane |
C5H12O | |
MFCD00008812 | |
tert-butyl methyl ether, methyl tert-butyl ether, mtbe, methyl t-butyl ether, t-butyl methyl ether, methyl tertiary-butyl ether, propane, 2-methoxy-2-methyl, methyl-tert-butyl ether, methyl-t-butyl ether, 2-methyl-2-methoxypropane | |
CHEBI:27642 | |
CC(C)(C)OC |
Spécification
-110.0°C | |
White to Yellow | |
54.0°C to 56.0°C | |
Authentic | |
1.3675 to 1.3695 | |
0.7404 | |
(CH3)3COCH3 | |
0.36 mPa.s (20°C) | |
0.01% Max. (Coulometric) | |
15, 6106 | |
Solubility in water: 51g/L (20°C). Other solubilities: miscible with most organic solvents, unstable in acid solutions, soly of water in methyl-t-butyl ether : 1,5g/100g | |
CC(C)(C)OC | |
88.15 | |
CHEBI:27642 | |
99% | |
tert-Butyl methyl ether, 99% |
1634-04-4 | |
0.7404g/mL | |
−28°C | |
Powder or Needles | |
99% | |
C5H12O | |
MFCD00008812 | |
01, 381 | |
04,333 | |
tert-butyl methyl ether, methyl tert-butyl ether, mtbe, methyl t-butyl ether, t-butyl methyl ether, methyl tertiary-butyl ether, propane, 2-methoxy-2-methyl, methyl-tert-butyl ether, methyl-t-butyl ether, 2-methyl-2-methoxypropane | |
BZLVMXJERCGZMT-UHFFFAOYSA-N | |
2-methoxy-2-methylpropane | |
15413 | |
88.15 | |
Pure |
Sécurité et traitement
GHS P Statement
Causes skin irritation.
Highly flammable liquid and vapor.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
- tert-Butyl methyl ether
Mention d'avertissement
- Danger
Catégorie de danger
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
Mention de danger
- H225-Highly flammable liquid and vapour.
- H315-Causes skin irritation.
Conseils de prudence
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
RUO – Research Use Only
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.