missing translation for 'onlineSavingsMsg'
Learn More
tert-Butyl methyl ether, ACS reagent, Thermo Scientific Chemicals Product Code.: 11974701

tert-Butyl methyl ether, ACS reagent, Thermo Scientific Chemicals

Product Code. 11974701
1 L
Click to view available options
Quantity:
1 L
2.5 L
This item is not returnable. View return policy

Product Code. 11974701

Brand: Thermo Scientific Acros 447470010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1634-04-4
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
InChI Key BZLVMXJERCGZMT-UHFFFAOYSA-N
IUPAC Name 2-methoxy-2-methylpropane
SMILES COC(C)(C)C

Specifications

Melting Point -110.0°C
Density 0.7404 g/mL
Boiling Point 54.0°C to 56.0°C
Quantity 1 L
Flash Point -28°C
Packaging Glass Bottle
Refractive Index 1.3675 to 1.3695
Specific Gravity 0.7404
Linear Formula (CH3)3COCH3
Viscosity 0.36 mPa.s (20°C)
Merck Index 15, 6106
Solubility Information Solubility aq. soln.: 51 g/L (20°C). Other solubilities: miscible with most organic solvents, unstable in acid solutions, soly of water in methyl-t-butyl ether : 1,5 g/100 g
Formula Weight 88.15
Grade ACS Reagent
Chemical Name or Material tert-Butyl methyl ether
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466324 Certificate TypeCertificate of Analysis Date19/09/2024 Catalog Number447470010,447470025
Lot NumberA0462725 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number447470010,447470025
Lot NumberA0454029 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number447470010,447470025
Lot NumberA0449004 Certificate TypeCertificate of Analysis Date05/01/2023 Catalog Number447470010,447470025
Lot NumberA0445948 Certificate TypeCertificate of Analysis Date13/09/2022 Catalog Number447470010,447470025
compliance-icons compliance-icons
Product Identifier
  • tert-Butyl methyl ether
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
Supplemental information
  • MIXTURE LIST-Contains : Methyl tert-butyl ether

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
tert-Butyl methyl ether, ACS reagent, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.