missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl methyl ether, for analysis, Thermo Scientific Chemicals

58.79 € - 113.57 €

Chemical Identifiers

CAS 1634-04-4
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00008812
InChI Key BZLVMXJERCGZMT-UHFFFAOYSA-N
Synonym tert-butyl methyl ether, methyl tert-butyl ether, mtbe, methyl t-butyl ether, t-butyl methyl ether, methyl tertiary-butyl ether, propane, 2-methoxy-2-methyl, methyl-tert-butyl ether, methyl-t-butyl ether, 2-methyl-2-methoxypropane
PubChem CID 15413
ChEBI CHEBI:27642
IUPAC Name 2-methoxy-2-methylpropane
SMILES CC(C)(C)OC
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10197343
View Documents
Thermo Scientific Acros
390480010
1 L Glass Bottle
58.79 €
1 L
Please to purchase this item. Need a web account? Register with us today!
10433735
View Documents
Thermo Scientific Acros
390480025
2.5 L Glass Bottle
113.57 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1634-04-4
88.15
BZLVMXJERCGZMT-UHFFFAOYSA-N
15413
2-methoxy-2-methylpropane
C5H12O
MFCD00008812
tert-butyl methyl ether, methyl tert-butyl ether, mtbe, methyl t-butyl ether, t-butyl methyl ether, methyl tertiary-butyl ether, propane, 2-methoxy-2-methyl, methyl-tert-butyl ether, methyl-t-butyl ether, 2-methyl-2-methoxypropane
CHEBI:27642
CC(C)(C)OC

Specifications

-110.0°C
Colorless
0.7404g/mL
−28°C
Liquid
0.005% max.
C5H12O
MFCD00008812
0.05% max.
tert-butyl methyl ether, methyl tert-butyl ether, mtbe, methyl t-butyl ether, t-butyl methyl ether, methyl tertiary-butyl ether, propane, 2-methoxy-2-methyl, methyl-tert-butyl ether, methyl-t-butyl ether, 2-methyl-2-methoxypropane
BZLVMXJERCGZMT-UHFFFAOYSA-N
2-methoxy-2-methylpropane
15413
88.15
Analytical
1634-04-4
0.0005 meq/g max.
54.0°C to 56.0°C
Authentic
1.3675 to 1.3695
0.7404
(CH3)3COCH3
0.36 mPa.s (20°C)
15, 6106
Solubility in water: 51g/L (20°C). Other solubilities: miscible with most organic solvents, unstable in acid solutions, soly of water in methyl-t-butyl ether : 1,5g/100g
CC(C)(C)OC
88.15
CHEBI:27642
≥99.5%
tert-Butyl methyl ether
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Highly flammable liquid and vapor.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Ground/Bond container and receiving equipment.

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 216-653-1

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.