missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butyl methyl ether CHROMASOLV™, for HPLC, ≥99.8%, Honeywell Riedel-de Haën™
Click to view available options
Quantity:
100 mL
1 L
2 L
2.5 L
4 x 2.5 L
6 x 1 L
Description
- For HPLC and organic synthesis
- High-purity, multipurpose solvents
- High lot-to-lot consistency

Specifications
Specifications
Density | 0.74g/mL at 25°C |
Boiling Point | 55°C to 56°C |
Quantity | 2.5 L |
Refractive Index | n20/D 1.369 |
Linear Formula | (CH3)3COCH3 |
Beilstein | 1730942 |
Synonym | tert-butyl methyl ether, methyl tert-butyl ether, mtbe, methyl t-butyl ether, t-butyl methyl ether, methyl tertiary-butyl ether, propane, 2-methoxy-2-methyl, methyl-tert-butyl ether, methyl-t-butyl ether, 2-methyl-2-methoxypropane |
SMILES | CC(C)(C)OC |
Formula Weight | 88.15g/mol |
Percent Purity | ≥99.8% |
Show More |
Safety and Handling


Product Identifier
- tert-Butyl methyl ether
- Danger
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- H225-Highly flammable liquid and vapour.
- H315-Causes skin irritation.
- P243-Take precautionary measures against static discharge.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
tert-Butyl methyl ether CHROMASOLV™, for HPLC, ≥99.8%, Honeywell Riedel-de Haën™ >
Spot an opportunity for improvement?Share a Content Correction