missing translation for 'onlineSavingsMsg'
Learn More
tert-Butyl N-[(1-benzyl-4-piperidinyl)methyl]carbamate, ≥95%, Thermo Scientific™ Product Code.: 10126772

tert-Butyl N-[(1-benzyl-4-piperidinyl)methyl]carbamate, ≥95%, Thermo Scientific™

Product Code. 10126772
10 g, Amber Glass Bottle
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 10126772

Brand: Thermo Scientific Maybridge MO00974EA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 173340-23-3
Molecular Formula C18H28N2O2
Molecular Weight (g/mol) 304.434
MDL Number MFCD04123925
InChI Key SOXSDIKJHXTJEP-UHFFFAOYSA-N
Synonym tert-butyl 1-benzylpiperidin-4-yl methyl carbamate, tert-butyl 1-benzylpiperidin-4-yl methylcarbamate, tert-butyl n-1-benzyl-4-piperidinyl methyl carbamate, tert-butyl n-1-benzylpiperidin-4-yl methyl carbamate, maybridge3_004457, 1-benzyl-4-tertbutyloxycarbonylaminomethylpiperidine, 1-benzyl-4-piperidinylmethyl carbamic acid tert-butyl ester, 1-benzylpiperidin-4-yl methylcarbamic acid tert-butyl ester, carbamic acid,n-1-phenylmethyl-4-piperidinyl methyl-, 1,1-dimethylethyl ester
PubChem CID 2824066
IUPAC Name tert-butyl N-[(1-benzylpiperidin-4-yl)methyl]carbamate
SMILES CC(C)(C)OC(=O)NCC1CCN(CC1)CC2=CC=CC=C2

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 304.43
Percent Purity 95%
Chemical Name or Material tert-Butyl N-[(1-benzyl-4-piperidinyl)methyl]carbamate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
tert-Butyl N-[(1-benzyl-4-piperidinyl)methyl]carbamate, ≥95%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.