missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl N-(2-cyanoethyl)carbamate, 97%, Thermo Scientific™

Product Code. 10079481
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 53588-95-7
Molecular Formula C8H14N2O2
Molecular Weight (g/mol) 170.21
MDL Number MFCD02180880
InChI Key NORLFIHQJFOIGS-UHFFFAOYSA-N
Synonym tert-butyl n-2-cyanoethyl carbamate, tert-butyl 2-cyanoethyl carbamate, n-boc-2-cyano-ethylamine, 2-cyano-ethyl-carbamic acid tert-butyl ester, t-butyl n-2-cyanoethyl carbamate, tert-butoxy-n-2-cyanoethyl carboxamide, 1,1-dimethylethyl 2-cyanoethyl carbamate, 3-tert-butoxycarbonyl amino propanenitrile, carbamicacid,n-2-cyanoethyl-,1,1-dimethylethylester
PubChem CID 2735664
IUPAC Name tert-butyl N-(2-cyanoethyl)carbamate
SMILES CC(C)(C)OC(=O)NCCC#N

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Linear Formula (CH3)3COC(O)NH(CH2)2CN
Packaging Amber Glass Bottle
Formula Weight 170.21
Percent Purity 97%
Quantity 1 g
Chemical Name or Material Tert-butyl N-(2-cyanoethyl)carbamate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'