missing translation for 'onlineSavingsMsg'
Learn More
tert-Butyl N-(3-amino-3-thioxopropyl)carbamate, 97%, Thermo Scientific™ Product Code.: 10437832

tert-Butyl N-(3-amino-3-thioxopropyl)carbamate, 97%, Thermo Scientific™

Product Code. 10437832
1 g, Amber Glass Bottle
Click to view available options
Quantity:
1 g
5 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 10437832

Brand: Thermo Scientific Maybridge SPB08402DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 77152-97-7
Molecular Formula C8H16N2O2S
Molecular Weight (g/mol) 204.288
MDL Number MFCD02180883
InChI Key OBDMXQCRRWGEQM-UHFFFAOYSA-N
Synonym tert-butyl n-3-amino-3-thioxopropyl carbamate, tert-butyl 3-amino-3-thioxopropylcarbamate, tert-butyl n-2-carbamothioylethyl carbamate, tert-butyl 3-amino-3-thioxopropyl carbamate, 3-tert-butoxycarbonyl amino propanethioamide, tert-butyl 3-amino-3-thioxoprop-1-yl carbamate, 2-thiocarbamoylethyl carbamic acid tert-butyl ester, carbamic acid,n-3-amino-3-thioxopropyl-, 1,1-dimethylethyl ester, tert-butyl 2-thiocarbamoylethylcarbamate, 3-tert-butoxycarbonylamino propanethioamide
PubChem CID 2735653
IUPAC Name tert-butyl N-(3-amino-3-sulfanylidenepropyl)carbamate
SMILES CC(C)(C)OC(=O)NCCC(=S)N

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula (CH3)3COC(O)NH(CH2)2C(S)NH2
Quantity 1 g
Formula Weight 204.29
Percent Purity 97%
Chemical Name or Material Tert-butyl N-(3-amino-3-thioxopropyl)carbamate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
tert-Butyl N-(3-amino-3-thioxopropyl)carbamate, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.