missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl N-[4-(aminomethyl)phenyl]carbamate, 97%, Thermo Scientific™

Product Code. 10136942
Click to view available options
Quantity:
250 mg
1 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 220298-96-4
Molecular Formula C12H18N2O2
Molecular Weight (g/mol) 222.29
MDL Number MFCD02183573
InChI Key URXUHALBOWYXJZ-UHFFFAOYSA-N
Synonym tert-butyl n-4-aminomethyl phenyl carbamate, 4-aminomethyl-1-n-boc-aniline, tert-butyl 4-aminomethyl phenyl carbamate, 4-boc-amino benzylamine, 4-aminomethy-1-n-boc-aniline, 4-n-boc-amino benzylamine, 4-tert-butoxycarbonylamino benzylamine, 4-aminomethyl-n-boc-aniline, tert-butyl-n-4-aminomethyl phenyl carbamate
PubChem CID 2794659
IUPAC Name tert-butyl N-[4-(aminomethyl)phenyl]carbamate
SMILES CC(C)(C)OC(=O)NC1=CC=C(CN)C=C1

Specifications

CAS Min % 97.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 222.29
Percent Purity 97%
Chemical Name or Material tert-Butyl N-[4-(aminomethyl)phenyl]carbamate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'