missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl N-(4-piperidinylmethyl)carbamate, 97%, Thermo Scientific™

Product Code. 10744023
1 g
Click to view available options
Quantity:
250 mg
1 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 135632-53-0
Molecular Formula C11H22N2O2
Molecular Weight (g/mol) 214.309
InChI Key VHYXAWLOJGIJPC-UHFFFAOYSA-N
Synonym tert-butyl piperidin-4-ylmethyl carbamate, 4-boc-aminomethyl piperidine, 4-tert-butoxycarbonylaminomethyl piperidine, 4-n-boc-aminomethyl piperidine, 4-boc-aminomethyl-piperidine, tert-butyl n-piperidin-4-ylmethyl carbamate, tert-butyl piperidin-4-ylmethylcarbamate, boc-4-aminomethyl piperidine
PubChem CID 723429
IUPAC Name tert-butyl N-(piperidin-4-ylmethyl)carbamate
SMILES CC(C)(C)OC(=O)NCC1CCNCC1

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Formula Weight 214.31
Percent Purity 95%
Quantity 1 g
Chemical Name or Material tert-Butyl N-(4-piperidinylmethyl)carbamate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
tert-Butyl N-(4-piperidinylmethyl)carbamate, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.