missing translation for 'onlineSavingsMsg'
Learn More
tert-Butyl N-[(5-bromo-2-thienyl)methyl]carbamate, 97%, Thermo Scientific™ Product Code.: 10681873

tert-Butyl N-[(5-bromo-2-thienyl)methyl]carbamate, 97%, Thermo Scientific™

Product Code. 10681873
1 g, Amber Glass Bottle
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 10681873

Brand: Thermo Scientific Maybridge MO00599DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 215183-27-0
Molecular Formula C10H14BrNO2S
Molecular Weight (g/mol) 292.191
MDL Number MFCD03659715
InChI Key CVNITIPQHIGCKI-UHFFFAOYSA-N
Synonym tert-butyl 5-bromothiophen-2-yl methyl carbamate, tert-butyl n-5-bromo-2-thienyl methyl carbamate, 5-bromo-thiophen-2-ylmethyl-carbamic acid tert-butyl ester, tert-butyl 5-bromothiophen-2-yl methylcarbamate, tert-butyl n-5-bromothiophen-2-yl methyl carbamate, n-boc-5-bromothiophen-2-methanamine, 2-boc-amino methyl-5-bromothiophene, 2-bromo-5-tert-butoxycarbonylaminomethylthiophene
PubChem CID 2779810
IUPAC Name tert-butyl N-[(5-bromothiophen-2-yl)methyl]carbamate
SMILES CC(C)(C)OC(=O)NCC1=CC=C(S1)Br

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 292.2
Percent Purity 97%
Chemical Name or Material Tert-butyl N-[(5-bromo-2-thienyl)methyl]carbamate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
tert-Butyl N-[(5-bromo-2-thienyl)methyl]carbamate, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.