missing translation for 'onlineSavingsMsg'
Learn More
tert-Butyl (S)-(-)-4-formyl-2,2-dimethyl-3-oxazolidinecarboxylate, 95%, Thermo Scientific Chemicals Product Code.: 10404442

tert-Butyl (S)-(-)-4-formyl-2,2-dimethyl-3-oxazolidinecarboxylate, 95%, Thermo Scientific Chemicals

Product Code. 10404442
1 g
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 10404442

Brand: Thermo Scientific Acros 386570010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102308-32-7
Molecular Formula C11H19NO4
Molecular Weight (g/mol) 229.28
MDL Number MFCD00209557
InChI Key PNJXYVJNOCLJLJ-MRVPVSSYSA-N
Synonym s---3-boc-2,2-dimethyloxazolidine-4-carboxaldehyde, tert-butyl s---4-formyl-2,2-dimethyl-3-oxazolidinecarboxylate, tert-butyl 4s-4-formyl-2,2-dimethyl-1,3-oxazolidine-3-carboxylate, unii-41aj913hqi, s-tert-butyl 4-formyl-2,2-dimethyloxazolidine-3-carboxylate, 1,1-dimethylethyl-s-4-formyl-2,2-dimethyl-3-oxazolidine carboxylate, s---3-tert-butoxycarbonyl-4-formyl-2,2-dimethyl-1,3-oxazolidine, s-4-formyl-2,2-dimethyl-oxazolidine-3-carboxylic acid tert-butyl ester, 3-oxazolidinecarboxylic acid, 4-formyl-2,2-dimethyl-, 1,1-dimethylethyl ester, 4s, s---3-boc-4-formyl-2,2-dimethyl-1,3-oxazolidine
PubChem CID 179824
IUPAC Name tert-butyl (4S)-4-formyl-2,2-dimethyl-1,3-oxazolidine-3-carboxylate
SMILES CC1(N(C(CO1)C=O)C(=O)OC(C)(C)C)C

Specifications

Color Colorless to Yellow
Density 1.0600g/mL
Boiling Point 83°C to 88°C (1.0 mmHg)
Flash Point 108°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4450 to 1.4490 (20°C, 589nm)
Quantity 1 g
Merck Index 15,4404
Specific Gravity 1.06
Specific Rotation -85° to -95° (20°C, 589nm) (c=1, CHCl3)
Formula Weight 229.28
Percent Purity 95%
Physical Form Oil
Chemical Name or Material tert-Butyl (S)-(-)-4-formyl-2, 2-dimethyl-3-oxazolidinecarboxylate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0448676 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number386570010
Lot NumberA0448676 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number386570010,386572500
Lot NumberA0389200 Certificate TypeCertificate of Analysis Date28/06/2021 Catalog Number386570010,386572500
compliance-icons
Product Identifier
  • tert-Butyl (S)-(-)-4-formyl-2,2-dimethyl-3-oxazolidinecarboxylate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
tert-Butyl (S)-(-)-4-formyl-2,2-dimethyl-3-oxazolidinecarboxylate, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.