missing translation for 'onlineSavingsMsg'
Learn More

tert-Butylacetyl chloride, 98+%, Thermo Scientific Chemicals

21.41 € - 671.34 €

Chemical Identifiers

CAS 7065-46-5
Molecular Formula C6H11ClO
Molecular Weight (g/mol) 134.603
MDL Number MFCD00000737
InChI Key BUTKIHRNYUEGKB-UHFFFAOYSA-N
Synonym 3,3-dimethylbutyryl chloride, tert-butylacetyl chloride, t-butylacetyl chloride, butanoyl chloride, 3,3-dimethyl, 3,3-dimethyl butyryl chloride, tert-butyl acetyl chloride, neohexanoyl chloride, t-butylacetylchloride, tertbutylacetylchloride, pubchem2093
PubChem CID 81514
IUPAC Name 3,3-dimethylbutanoyl chloride
SMILES CC(C)(C)CC(=O)Cl
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11453950
missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
A10162.06
5 g
21.41 €
5g
Please to purchase this item. Need a web account? Register with us today!
11459442
missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
A10162.14
25 g
48.84 €
25g
Please to purchase this item. Need a web account? Register with us today!
11469442
missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
A10162.22
100 g
152.76 €
100g
Please to purchase this item. Need a web account? Register with us today!
11479442
missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
A10162.36
500 g
671.34 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

tert-Butylacetyl chloride is used an acyl chloride compound for proteomics research. It is also used as intermediate for important acylation reagents agrochemical and pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butylacetyl chloride is used an acyl chloride compound for proteomics research. It is also used as intermediate for important acylation reagents agrochemical and pharmaceutical.

Solubility
Reacts with water.

Notes
Moisture Sensitive Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

7065-46-5
134.603
BUTKIHRNYUEGKB-UHFFFAOYSA-N
81514
CC(C)(C)CC(=O)Cl
C6H11ClO
MFCD00000737
3,3-dimethylbutyryl chloride, tert-butylacetyl chloride, t-butylacetyl chloride, butanoyl chloride, 3,3-dimethyl, 3,3-dimethyl butyryl chloride, tert-butyl acetyl chloride, neohexanoyl chloride, t-butylacetylchloride, tertbutylacetylchloride, pubchem2093
3,3-dimethylbutanoyl chloride

Specifications

7065-46-5
21°C (69°F)
C6H11ClO
UN2924
3,3-dimethylbutyryl chloride, tert-butylacetyl chloride, t-butylacetyl chloride, butanoyl chloride, 3,3-dimethyl, 3,3-dimethyl butyryl chloride, tert-butyl acetyl chloride, neohexanoyl chloride, t-butylacetylchloride, tertbutylacetylchloride, pubchem2093
CC(C)(C)CC(=O)Cl
134.603
134.61
tert-Butylacetyl chloride
128°C to 130°C
Acrid
MFCD00000737
1740421
BUTKIHRNYUEGKB-UHFFFAOYSA-N
3,3-dimethylbutanoyl chloride
81514
≥98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.

missing translation for 'tsca' : TSCA

Recommended Storage : 2°C to 8°C

SDS
Documents

Documents

Certificates

RUO – Research Use Only