missing translation for 'onlineSavingsMsg'
Learn More

tert-Butylacetyl chloride, 99%, Thermo Scientific Chemicals

31.36 € - 334.67 €

Chemical Identifiers

CAS 7065-46-5
Molecular Formula C6H11ClO
Molecular Weight (g/mol) 134.61
MDL Number MFCD00000737
InChI Key BUTKIHRNYUEGKB-UHFFFAOYSA-N
Synonym 3,3-dimethylbutyryl chloride, tert-butylacetyl chloride, t-butylacetyl chloride, butanoyl chloride, 3,3-dimethyl, 3,3-dimethyl butyryl chloride, tert-butyl acetyl chloride, neohexanoyl chloride, t-butylacetylchloride, tertbutylacetylchloride, pubchem2093
PubChem CID 81514
IUPAC Name 3,3-dimethylbutanoyl chloride
SMILES CC(C)(C)CC(=O)Cl
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10094093
View Documents
Thermo Scientific Acros
107790100
10 mL Glass bottle
31.36 €
10mL
Please to purchase this item. Need a web account? Register with us today!
10705751
View Documents
Thermo Scientific Acros
107790500
50 mL Glass bottle
81.00 €
50mL
Please to purchase this item. Need a web account? Register with us today!
10599990
View Documents
Thermo Scientific Acros
107792500
250 mL Glass bottle
334.67 €
250mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7065-46-5
134.61
BUTKIHRNYUEGKB-UHFFFAOYSA-N
81514
CC(C)(C)CC(=O)Cl
C6H11ClO
MFCD00000737
3,3-dimethylbutyryl chloride, tert-butylacetyl chloride, t-butylacetyl chloride, butanoyl chloride, 3,3-dimethyl, 3,3-dimethyl butyryl chloride, tert-butyl acetyl chloride, neohexanoyl chloride, t-butylacetylchloride, tertbutylacetylchloride, pubchem2093
3,3-dimethylbutanoyl chloride

Specifications

7065-46-5
0.9600g/mL
21°C
99%
(CH3)3CCH2COCl
MFCD00000737
3,3-dimethylbutyryl chloride, tert-butylacetyl chloride, t-butylacetyl chloride, butanoyl chloride, 3,3-dimethyl, 3,3-dimethyl butyryl chloride, tert-butyl acetyl chloride, neohexanoyl chloride, t-butylacetylchloride, tertbutylacetylchloride, pubchem2093
BUTKIHRNYUEGKB-UHFFFAOYSA-N
3,3-dimethylbutanoyl chloride
81514
99%
tert-Butylacetyl chloride, 99%
Colorless to Yellow
127.0°C to 129.0°C
Authentic
C6H11ClO
1.4210 to 1.4230
0.96
Solubility in water: reacts. Other solubilities: soluble in ethanol
CC(C)(C)CC(=O)Cl
134.61
134.61
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.

GHS P Statement
Wear eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 230-356-4

Recommended Storage : Refrigerator +4°C

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.