Learn More
tert-Butylamine, 98%, Thermo Scientific Chemicals
Description
tert-butylamine is used as a precursor for the preparation of rubber accelerators, pharmaceuticals and dyes. It is employed in synthesis of N-tert-butyl-acetamide, isoquinolines, pyridines, decumbenine, sulfontes, sulfonamides and methyl beta-ketophosphonate. It is also used in the study the effect of amines in influencing atmospheric sulfuric acid - water nucleation, hydroamination of terminal alkynes and measuring the surface acidity of solid catalyst. It acts as an indicator molecule in the study of NMR -based pH determination method.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
tert-butylamine is used as a precursor for the preparation of rubber accelerators, pharmaceuticals and dyes. It is employed in synthesis of N-tert-butyl-acetamide, isoquinolines, pyridines, decumbenine, sulfontes, sulfonamides and methyl beta-ketophosphonate. It is also used in the study the effect of amines in influencing atmospheric sulfuric acid - water nucleation, hydroamination of terminal alkynes and measuring the surface acidity of solid catalyst. It acts as an indicator molecule in the study of NMR -based pH determination method.
Solubility
Miscible with water, alcohol, ether, chloroform and organic solvents.
Notes
Air sensitive. Incompatible with strong oxidizing agents, strong acids and acid anhydrides.
Specifications
Specifications
Melting Point | -67°C |
Density | 0.696 |
Boiling Point | 44°C to 46°C |
Flash Point | −38°C (−36°F) |
Odor | Amine-like |
Refractive Index | 1.378 |
Quantity | 2500 mL |
UN Number | UN2733 |
Beilstein | 605267 |
Sensitivity | Air Sensitive |
Show More |
RUO – Research Use Only