missing translation for 'onlineSavingsMsg'
Learn More
tert-Butylamine, 99%, AcroSeal™, Thermo Scientific Chemicals Product Code.: 10009243

tert-Butylamine, 99%, AcroSeal™, Thermo Scientific Chemicals

Product Code. 10009243
100 mL, AcroSeal™ Glass bottle
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Product Code. 10009243

Brand: Thermo Scientific Acros 427241000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-64-9
Molecular Formula C4H11N
Molecular Weight (g/mol) 73.13
MDL Number MFCD00008050
InChI Key YBRBMKDOPFTVDT-UHFFFAOYSA-N
Synonym tert-butylamine, erbumine, 2-amino-2-methylpropane, t-butylamine, 2-propanamine, 2-methyl, trimethylaminomethane, 1,1-dimethylethylamine, 2-aminoisobutane, 1,1-dimethylethanamine, butylamine, tert
PubChem CID 6385
ChEBI CHEBI:44639
IUPAC Name 2-methylpropan-2-amine
SMILES CC(C)(C)N

Specifications

Melting Point -67.0°C
Density 0.6900g/mL
Boiling Point 46.0°C
Flash Point −38°C
Assay Percent Range 98.5% min. (GC)
Packaging AcroSeal™ Glass bottle
Linear Formula (CH3)3CNH2
Refractive Index 1.3770 to 1.3790
Quantity 100 mL
Merck Index 15, 1547
Specific Gravity 0.69
Solubility Information Solubility in water: 1000g/L (25°C). Other solubilities: miscible with alcohol,soluble in common organic solvents
Viscosity 0.5 mPa.s (20°C)
Formula Weight 73.13
Percent Purity 99%
Chemical Name or Material tert-Butylamine
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0457253 Certificate TypeCertificate of Analysis Date23/10/2023 Catalog Number427248000,427241000
Lot NumberA0446072 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number427248000,427241000
Lot NumberA0426334 Certificate TypeCertificate of Analysis Date12/01/2023 Catalog Number427248000,427241000
Lot NumberA0395834 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number427248000,427241000
Lot NumberA0403009 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog Number427248000,427241000
compliance-icons compliance-icons compliance-icons
Product Identifier
  • tert-Butylamine
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
  • Acute toxicity Category 4
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H331-Toxic if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
tert-Butylamine, 99%, AcroSeal™, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.