missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butylbenzene, 99%
Click to view available options
Quantity:
250 mL
1000 mL
Unit Size:
1000mL
250mL
Description
tert-Butylbenzene is an useful intermediate for manufacturing agrochemicals and pharmaceuticals. tert-Butyl Toluene is used as a solvent for resins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
tert-Butylbenzene is an useful intermediate for manufacturing agrochemicals and pharmaceuticals. tert-Butyl Toluene is used as a solvent for resins.
Solubility
Insoluble in water(0.03g/100 mL at 20°C) .
Notes
Keep container tightly closed in a cool, dry and well-ventilated place. Store away from strong oxidizing agents, acids.
tert-Butylbenzene is an useful intermediate for manufacturing agrochemicals and pharmaceuticals. tert-Butyl Toluene is used as a solvent for resins.
Solubility
Insoluble in water(0.03g/100 mL at 20°C) .
Notes
Keep container tightly closed in a cool, dry and well-ventilated place. Store away from strong oxidizing agents, acids.
Specifications
Specifica
| Melting Point | -58°C |
| Density | 0.867 |
| Boiling Point | 168°C to 169°C |
| Flash Point | 34°C (93°F) |
| Odor | Characteristic |
| Refractive Index | 1.492 |
| Quantity | 250 mL |
| UN Number | UN2709 |
| Beilstein | 1421537 |
| Merck Index | 14,1551 |
| Vedi altri risultati |
Sicurezza e movimentazione
Identificativo prodotto
- tert-Butylbenzene
- Warning
- Flammable liquid Category 3
- H226-Flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- MIXTURE LIST-Contains : tert-Butylbenzene
RUO – Research Use Only
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto