missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyldimethylchlorosilane, 97%, Thermo Scientific Chemicals

Product Code. 11484364
Click to view available options
Quantity:
5 g
25 g
100 g

tert-Butyldimethylchlorosilane is used to protect hydroxyl group in organic synthesis. It finds application in the synthesis of prostaglandin. It is also used as an auxiliary material for hypolipaemics such as lovastatin and simvastatin. It plays an important role in the preparation of isoxazolines N-oxides from alpha-bromonitroalkanes. It acts as a versatile protecting reagent for amines, amides and alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyldimethylchlorosilane is used to protect hydroxyl group in organic synthesis. It finds application in the synthesis of prostaglandin. It is also used as an auxiliary material for hypolipaemics such as lovastatin and simvastatin. It plays an important role in the preparation of isoxazolines N-oxides from alpha-bromonitroalkanes. It acts as a versatile protecting reagent for amines, amides and alcohols.

Solubility
Soluble in chloroform and ethyl acetate. Insoluble in water.

Notes
Moisture sensitive. Store in cool place. Incompatible with bases, metals, oxidizing agents and alcohols.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18162-48-6
Molecular Formula C6H15ClSi
Molecular Weight (g/mol) 150.721
MDL Number MFCD00000501
InChI Key BCNZYOJHNLTNEZ-UHFFFAOYSA-N
Synonym tert-butyldimethylsilyl chloride, tert-butyldimethylchlorosilane, t-butyldimethylchlorosilane, tert-butylchlorodimethylsilane, tbdms chloride, tert-butyl chloro dimethylsilane, chloro-tert-butyldimethylsilane, silane, chloro 1,1-dimethylethyl dimethyl, tbscl, t-butyldimethylsilyl chloride
PubChem CID 28928
ChEBI CHEBI:85071
IUPAC Name tert-butyl-chloro-dimethylsilane
SMILES CC(C)(C)[Si](C)(C)Cl

Specifications

Boiling Point 124°C to 126°C
Melting Point 86°C to 89°C
Quantity 25 g
Linear Formula (CH3)3CSi(CH3)2Cl
UN Number UN2925
Beilstein 505999
Flash Point 22°C (72°F)
Solubility Information Soluble in chloroform and ethyl acetate. Insoluble in water.
Formula Weight 150.73
Percent Purity 97%
Odor Pungent
Sensitivity Moisture sensitive
Density 0.81
Chemical Name or Material tert-Butyldimethylchlorosilane
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • tert-Butyldimethylchlorosilane
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H228-Flammable solid.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : chloro(1,1-dimethylethyl)dimethyl-silane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.