missing translation for 'onlineSavingsMsg'
Learn More
tert-Butylmagnesium chloride, 1.7M solution in diethyl ether, AcroSeal™, Thermo Scientific Chemicals Product Code.: 10688123

tert-Butylmagnesium chloride, 1.7M solution in diethyl ether, AcroSeal™, Thermo Scientific Chemicals

Product Code. 10688123
100 mL, AcroSeal™ Glass Bottle in Tin Can
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
AcroSeal™ Glass Bottle in Tin Can
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Product Code. 10688123

Brand: Thermo Scientific Acros 394301000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 677-22-5, 60-29-7
Molecular Formula C4H9ClMg
Molecular Weight (g/mol) 116.87
MDL Number MFCD00000465
InChI Key ZDRJSYVHDMFHSC-UHFFFAOYSA-M
Synonym tert-butylmagnesium chloride, tert-butylmagnesiumchloride, t-butylmagnesium chloride, t-butyl magnesium chloride, tert-butyl chloro magnesium, magnesium, chloro 1,1-dimethylethyl, tert-butylmagnesium chloride solution, 1.0 m in thf, tbumgcl, tert-bumgcl, t-bumgclShow MoreShow Less
PubChem CID 2724198
IUPAC Name magnesium;2-methylpropane;chloride
SMILES CC(C)(C)[Mg]Cl

Specifications

Density 0.8g/mL
Packaging AcroSeal™ Glass Bottle in Tin Can
Linear Formula (CH3)3CMgCl
Quantity 100 mL
Formula Weight 116.87
Chemical Name or Material tert-Butylmagnesium chloride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0436543 Certificate TypeCertificate of Analysis Date27/11/2023 Catalog Number394301000,394308000
Lot NumberA0452818 Certificate TypeCertificate of Analysis Date16/05/2023 Catalog Number394301000,394308000
Lot NumberA0403791 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number394301000,394308000
Lot NumberA0435017 Certificate TypeCertificate of Analysis Date11/11/2021 Catalog Number394301000,394308000
Lot NumberA0412010 Certificate TypeCertificate of Analysis Date10/09/2020 Catalog Number394301000,394308000
compliance-icons compliance-icons compliance-icons
Product Identifier
  • tert-Butylmagnesium chloride, solution in diethylether
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
  • Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H224-Extremely flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
tert-Butylmagnesium chloride, 1.7M solution in diethyl ether, AcroSeal™, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.