missing translation for 'onlineSavingsMsg'
Learn More

tert-Nitrosobutane dimer, 98+%

Product Code. 11340054
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 11340054

Brand: Thermo Scientific Alfa Aesar L00462.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6841-96-9 | C8H18N2O2 | 174.244 g/mol

tert-Nitrosobutane dimer is used as a spin trapping reagent in electron spin resonance studies. Spin trap reagent, Reactant involved in the synthesis of air-stable π-radical based on Corannulene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Nitrosobutane dimer is used as a spin trapping reagent in electron spin resonance studies. Spin trap reagent, Reactant involved in the synthesis of air-stable π-radical based on Corannulene.

Solubility
Soluble in 5% chloroform.

Notes
Light Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6841-96-9
Molecular Formula C8H18N2O2
Molecular Weight (g/mol) 174.244
MDL Number MFCD00002065
InChI Key PKKJRWITDTTZCL-UHFFFAOYSA-N
Synonym dimer, nitroso-t-butane, nitroso-tert-butane, tert-nitrosobutane dimer, 2-methyl-2-nitrosopropane, dimer, bis, 2-methyl-2-nitroso propane dimer, 1,1,1 inverted exclamation marka,1 inverted exclamation marka-tetramethylazoethane dioxide
PubChem CID 44134826
IUPAC Name 2-methyl-2-nitrosopropane
SMILES CC(C)(C)N=O.CC(C)(C)N=O

Specifications

Melting Point 88°C to 92°C (sublimation)
Quantity 1 g
Sensitivity Light sensitive
Solubility Information Soluble in 5% chloroform.
Formula Weight 174.24
Percent Purity ≥98%
Chemical Name or Material tert-Nitrosobutane dimer

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.