Learn More
Terumo™ Hypodermic Needles
Product Code. p-184970
missing translation for 'orderingAttributeHoverText'
Needle Gauge:
21 G
23 G
25 G
Length (English) Needle:
1 in.
Needle Point Style:
Thin Wall
Ultra-thin Wall
missing translation for 'unitSize'
Pack of 100
Product Code. 13144724
missing translation for 'mfr': Terumo™ TUAN2325R1
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Provides ultra sharp needles, which translate into greater comfort for the patient.
  • Double bevel design
  • Precision manufactured ultrathin wall
Specifications
Needle Gauge 23 G
Latex Free Yes
Length (English) Needle 1 in.
Product Type Double Bevel Design
Needle Point Style Ultra-thin Wall