Learn More
TES-1025, MedChemExpress
MedChemExpress TES-1025 is a potent and selective human α-amino-β-carboxymuconate-ε-semialdehyde decarboxylase (ACMSD) inhibitor with an IC50 of 13 nM.
Brand:  Medchem Express HY-111365
Encompass_Preferred
 View more versions of this product
Product Code. 16417157
  • 332.10 € / 5mg
Estimated Shipment: 27-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
TES-1025 is a potent and selective human α-amino-β-carboxymuconate-ε-semialdehyde decarboxylase (ACMSD) inhibitor with an IC50 of 13 nM.
Specifications
Specifications
TES-1025
Yellow
C18H13N3O3S2
DMSO : 100 mg/mL (260.80 mM; Need warming)
383.44
95.11%
Solid
1883602-21-8
Research
5 mg
O=C1C(C#N)=C(C2=CC=CS2)N=C(SCC3=CC=CC(CC(O)=O)=C3)N1
383.44
Research
Metabolism-protein/nucleotide metabolism