Learn More
Thermo Scientific Chemicals TES, 99%, for biochemistry
70.65 € - 131.66 €
Chemical Identifiers
CAS | 7365-44-8 |
---|---|
Molecular Formula | C6H15NO6S |
Molecular Weight (g/mol) | 229.247 |
MDL Number | MFCD00007532 |
InChI Key | JOCBASBOOFNAJA-UHFFFAOYSA-N |
Synonym | 2-[Tris(hydroxymethyl)methylamino]-1-ethanesulfonic acid |
PubChem CID | 81831 |
ChEBI | CHEBI:44356 |
IUPAC Name | 2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]ethanesulfonic acid |
SMILES | C(CS(=O)(=O)O)NC(CO)(CO)CO |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10358200
|
Thermo Scientific Chemicals
207510250 |
25 g | Glass bottle |
70.65 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10147753
|
Thermo Scientific Chemicals
207511000 |
100 g | Glass bottle |
131.66 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological bufferChemical Identifiers
7365-44-8 | |
229.247 | |
JOCBASBOOFNAJA-UHFFFAOYSA-N | |
81831 | |
2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]ethanesulfonic acid |
C6H15NO6S | |
MFCD00007532 | |
2-[Tris(hydroxymethyl)methylamino]-1-ethanesulfonic acid | |
CHEBI:44356 | |
C(CS(=O)(=O)O)NC(CO)(CO)CO |
Specifications
(0.1M aq. soln.), (0.1M aq. soln.) at 0nm, 0.02 max. at 280nm, 0.05 max. at 260nm | |
7365-44-8 | |
White | |
Crystalline Powder | |
Authentic | |
C6H15NO6S | |
MFCD00007532 | |
2-[Tris(hydroxymethyl)methylamino]-1-ethanesulfonic acid | |
JOCBASBOOFNAJA-UHFFFAOYSA-N | |
2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]ethanesulfonic acid | |
81831 | |
229.25 | |
Biochemical |
(0.1M aq. soln.) | |
223.0°C to 225.0°C | |
4 to 8 (0.1M soln. at 20°C) | |
99% | |
1% max. | |
(HOCH2)3CNHCH2CH2SO3H | |
15, 9319 | |
Solubility in water: soluble. | |
C(CS(=O)(=O)O)NC(CO)(CO)CO | |
229.247 | |
CHEBI:44356 | |
99% | |
TES, For biochemistry |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning
RUO – Research Use Only