missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IKA™ Test Tube Attachment for IKA Vortex Shaker
Attachment with customized array for holding reagent glasses on a vortexed.
42.50 € - 43.50 €
Specifications
Description | Test tube adapter attachment |
---|---|
Requires | VG 3.3 Universal Attachment |
Product Code | Brand | Holds | Product Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Product Type | Price | Quantity & Availability | ||||
10307662
|
IKA™
0003343900 |
18 x 10 mm Reagent Glasses |
42.50 €
Each |
Estimated Shipment: 26-02-2025
Log in to see stock. |
|||||
10367702
|
IKA™
0003344000 |
12 x 12 mm Reagent Glasses | Test Tube Adapter Attachment |
42.50 €
Each |
Estimated Shipment: 13-03-2025
Log in to see stock. |
||||
10656263
|
IKA™
0003344100 |
8 x 16 mm Reagent Glasses | Test Tube Adapter Attachment |
42.50 €
Each |
Estimated Shipment: 26-02-2025
Log in to see stock. |
||||
10414522
|
IKA™
0003344200 |
8 x 20 mm Reagent Glasses | Test Tube Adapter Attachment |
43.50 €
Each |
Estimated Shipment: 29-01-2025
Log in to see stock. |
||||
Description
- Requires VG 3.3 universal attachment
- Facilitates multi-tube vortexing
Specifications
Test tube adapter attachment | |
VG 3.3 Universal Attachment |