missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pinnacle Brush™ Test Tube Brushes with Cotton Yarn Tip
Features With cotton yarn tip. Pinnacle Brush™ Test Tube Brushes with Cotton Yarn Tip have a galvanised steel wire handle.
Brand: Pinnacle Brush™ T669
Description
- Cotton yarn tip
- Metal handle
Specifications
Test Tube Brush | |
Metal | |
25 mm | |
22 cm | |
15 cm | |
Brush Circumference: 11 cm |
Fiber | |
3.5 cm | |
Test tubes | |
7 cm | |
10 Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction