missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Test Tube Brushes with Nylon Head
Features fan shaped end. Pinnacle Brush™ Test Tube Brushes with Nylon Head thoroughly cleans test tubes.
Brand: PINNACLE Brush T664
Description
- With fan-shaped end
- Galvanised steel wire handle
Specifications
Test Tube Brush | |
Metal | |
Test tubes | |
6 cm | |
10 Pk. |
Nylon | |
1.6 cm | |
19 cm | |
13 cm | |
Brush Circumference: 5 cm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction