missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Test Tube Racks for MaxQ™ Shaker
Mix and match these shaker test tube racks, available in a variety of sizes and arrays, to suit individual needs in the laboratory.
Brand: Thermo Scientific™ 30183
This item is not returnable.
View return policy
Description
Mix and match Thermo Scientific™ MaxQ™ Shaker Test Tube Racks, available in a variety of sizes and arrays, to suit individual needs in the laboratory. All racks are compatible with MaxQ shaker platforms.
Racks are available in a multitude of sizes, arrays, and colors. Compatible with MaxQ shaker platforms.Specifications
Test Tube Rack | |
6 x 6 | |
14 to 16 mm Tubes, 15 mL Centrifuge Tubes | |
MaxQ™ Shaker Platform |
Orange; 14 to 16 mm; 6 x 6 array (fits 15 mL centrifuge tubes) | |
Orange | |
36 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction