missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Test Tube Racks for MaxQ™ Shaker

Mix and match these shaker test tube racks, available in a variety of sizes and arrays, to suit individual needs in the laboratory.

Brand:  Thermo Scientific™ 30189

 View more versions of this product

Product Code. 11496358

  • 382.91 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Mix and match Thermo Scientific™ MaxQ™ Shaker Test Tube Racks, available in a variety of sizes and arrays, to suit individual needs in the laboratory. All racks are compatible with MaxQ shaker platforms.

Racks are available in a multitude of sizes, arrays, and colors. Compatible with MaxQ shaker platforms.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Test Tube Rack
3 x 3
26 to 30 mm Tubes, 50 mL Centrifuge Tubes
MaxQ™ Shaker Platform
Green; 26 to 30 mm; 3 x 3 array (fits 50 mL centrifuge tubes)
Green
9
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.