Learn More
Fisherbrand™ Test Tubes, Borosilicate Glass 3.3, with Beaded Rim, Medium Wall
Test tubes with round bottom are universal cylindrical laboratory glassware that are used e.g. for chemical reactions, pretreatments or for storing small amounts of liquid.
25.35 € - 45.95 €
Product Code | Brand | Capacity (Metric) | Length (Metric) | Diameter (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Length (Metric) | Diameter (Metric) | Price | Quantity & Availability | |||
15852285
|
Fisherbrand™
90106153 |
3 mL | 75 mm | 10 mm |
40.35 €
Pack of 100 |
Estimated Shipment: 24-12-2024
Log in to see stock. |
|||
17921152
|
Fisherbrand™
90106192 |
5 mL | 75 mm | 12 mm |
25.35 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17951132
|
Fisherbrand™
90106193 |
13 mL | 100 mm | 16 mm |
42.05 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17941132
|
Fisherbrand™
90106194 |
17 mL | 125 mm | 16 mm |
43.05 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17901152
|
Fisherbrand™
90106196 |
21 mL | 150 mm | 16 mm |
44.50 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17961122
|
Fisherbrand™
90106186 |
21 mL | 160 mm | 16 mm |
45.00 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17931152
|
Fisherbrand™
90106197 |
27 mL | 150 mm | 18 mm |
45.95 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17911152
|
Fisherbrand™
90106198 |
53 mL | 150 mm | 24 mm |
35.50 €
Pack of 50 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17971122
|
Fisherbrand™
90106187 |
57 mL | 150 mm | 25 mm |
36.90 €
Pack of 50 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Transferring substances and liquids into other states of aggregation and thus initiating further reactions is very important for the daily work in the laboratory. A wide variety of vessels, apparatus and equipment are used for this work. Reaction controls, test series and preliminary samples are usually carried out in test tubes due to the small amounts of substance removed.
- Manufactured from borosilicate glass 3.3
- Thick-walled and therefore mechanically very resistant
- With round bottom