Learn More
Lenz Laborglasintrumente Test Tubes with NS Joint, with Conical Bottom
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 05705216
 View more versions of this product
Product Code. 10677780
  • 160.00 € / Pack of 10
Estimated Shipment: 04-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • With conical bottom
  • With socket NS 14/23
  • Available without stopper
  • With PE stopper or with hexagonal hollow glass stopper
  • Made of DURAN™ tubing
Specifications
Specifications
Hexagonal glass stopper
120 mm
17 mm