Learn More
Lenz Laborglasintrumente Test Tubes with NS Joint with Graduation
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
289.00 € - 294.00 €
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
10018289
|
Lenz Laborglasintrumente
05706003 |
25 mL |
294.00 €
Pack of 10 |
Estimated Shipment: 03-01-2025
Log in to see stock. |
|||||
10765262
|
Lenz Laborglasintrumente
05706002 |
20 mL |
289.00 €
Pack of 10 |
Estimated Shipment: 31-12-2024
Log in to see stock. |
|||||
Description
- With round bottom
- With NS socket
- With PE stopper
- With scale in high-contrast enamel
- Made of DURAN™ tubing
Specifications
Borosilicate Glass (3.3) |